N-(2,6-Diethylphenyl)maleimide 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(2,6-Diethylphenyl)-1H-pyrrole-2,5-dione
B20656.03
B20656.06
B20656.03EA
64.5
EUR
B20656.03
B20656.06
N-(2,6-Diethylphenyl)maleimide 97%
N-(2,6-Diethylphenyl)maleimide
Purity: 97%
CAS No.: 38167-72-5
Molecular Formula: C14H15NO2
Formula Weight: 229.28
MDL No.: MFCD00173826
Formel:
C₁₄H₁₅NO₂ Molecular Weight: 229,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173826 CAS-Nummer: 38167-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...