3,5-Dimethylbenzo[b]thiophene 97%

Lieferant: Thermo Scientific
Achtung

B20768.03 B20768.06
B20768.03EA 65.2 EUR
B20768.03 B20768.06
3,5-Dimethylbenzo[b]thiophene 97%
3,5-Dimethylbenzo[b]thiophene

Purity: 97% CAS No.: 1964-45-0 Molecular Formula: C10H10S Formula Weight: 162.25 Boiling Point: 62-64°/0.1mm MDL No.: MFCD00052744

Formel: C₁₀H₁₀S
Molecular Weight: 162,25 g/mol
Siedepunkt: 62…64 °C (0,1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052744
CAS-Nummer: 1964-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR