7-Nitro-1H-indole-2-carboxylic acid 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: CRT0044876

B20786.03 B20786.06
B20786.03EA 73.5 EUR
B20786.03 B20786.06
7-Nitro-1H-indole-2-carboxylic acid 96%
7-Nitro-1H-indole-2-carboxylic acid

Purity: 96% CAS No.: 6960-45-8 Molecular Formula: C9H6N2O4 Formula Weight: 206.16 Melting Point: 264-266° MDL No.: MFCD00044720

Formel: C₉H₆N₂O₄
Molecular Weight: 206,16 g/mol
Schmelzpunkt: 264…266 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044720
CAS-Nummer: 6960-45-8
EINECS: 230-154-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR