4-Methylumbelliferyl-β-D-galactopyranoside 98%
Lieferant: Thermo Scientific
Synonyme:
4-Methylumbelliferyl β-D-galactopyranoside, 4-Methyl-7-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one
B21075.01
B21075.03
B21075.01EA
30.4
EUR
B21075.01
B21075.03
4-Methylumbelliferyl-β-D-galactopyranoside 98%
4-Methylumbelliferyl-β-D-galactopyranoside
Purity: 98%
CAS No.: 6160-78-7
Molecular Formula: C16H18O8
Formula Weight: 338.32
Storage Temperature: -30°C to -10°C
Melting Point: ca 260° dec.
Beilstein Reference: 94673
Optical Rotation: -64° (c=1 in pyridine, 24h)
MDL No.: MFCD00036773
Formel:
C₁₆H₁₈O₈ Molecular Weight: 338,312 g/mol Schmelzpunkt: ∼ 260 °C (Zers.) |
MDL:
MFCD00036773 CAS-Nummer: 6160-78-7 EINECS: 228-185-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...