N-Fmoc-L-alanine monohydrate 98%
Lieferant: Thermo Scientific
Synonyme:
Fmoc-L-alanine monohydrate, N-(9-Fluorenylmethoxycarbonyl)-L-alanine monohydrate, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine monohydrate, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected monohydrate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine monohydrate, (-)-L-Alanine, N-FMOC protected monohydrate, (-)-FMOC-L-Ala-OH monohydrate, Fmoc-Ala-OH·H₂O, FMoc-ala-oh h2o, Fmoc-Ala-OH monohydrate
Purity: 98%
CAS No.: 207291-76-7
Molecular Formula: C18H17NO4.H2O
Formula Weight: 329.36 (311.34anhy)
Storage Temperature: 2°C to 8°C
Melting Point: ca 130°
Beilstein Reference: 2225975
Optical Rotation: -19° (c=1 in DMF)
MDL No.: MFCD00037139
Formel:
C₁₈H₁₇NO₄·H₂O Molecular Weight: 329,352 g/mol Schmelzpunkt: ∼ 130 °C |
MDL:
MFCD00037139 CAS-Nummer: 207291-76-7 EINECS: 252-660-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...