N-Vinyl-ε-caprolactam 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Vinylazepan-2-one
B21311.09
B21311.18
B21311.09EA
20.3
EUR
B21311.09
B21311.18
N-Vinyl-ε-caprolactam 99%
N-Vinyl-ε-caprolactam
Purity: 99%
CAS No.: 2235-00-9
Molecular Formula: C8H13NO
Formula Weight: 139.20
Flash Point: 101°(214°F)
Melting Point: 32-36°
Boiling Point: 128°/21mm
MDL No.: MFCD00080693
Formel:
C₈H₁₃NO Molecular Weight: 139,20 g/mol Siedepunkt: 128 °C (21 mmHg) Schmelzpunkt: 32…36 °C Dichte: 1,029 Flammpunkt: 101 °C (214 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00080693 CAS-Nummer: 2235-00-9 EINECS: 218-787-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...