(+/-)-ɑ-Bisabolol ≥96%

Lieferant: Thermo Scientific

Synonyme: rel-(R)-6-Methyl-2-((R)-4-methylcyclohex-3-en-1-yl)hept-5-en-2-ol, 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

B21457.09 B21457.18
B21457.09EA 56.3 EUR
B21457.09 B21457.18
(+/-)-ɑ-Bisabolol ≥96%
(+/-)-alpha-Bisabolol

Purity: 96% CAS No.: 515-69-5 Molecular Formula: C15H26O Formula Weight: 222.37 Flash Point: 135°(275°F) Refractive Index: 1.4950 Beilstein Reference: 5733954 MDL No.: MFCD03846910

Formel: C₁₅H₂₆O
Molecular Weight: 222,37 g/mol
Dichte: 0,93
Flammpunkt: 135 °C (275 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00129082
CAS-Nummer: 515-69-5
EINECS: 208-205-9
Merck Index: 14,1241

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR