Cyclohexyl hexanoate 99%

Lieferant: Thermo Scientific

B21547.14 B21547.22
B21547.14EA 72.5 EUR
B21547.14 B21547.22
Cyclohexyl hexanoate 99%
Cyclohexyl hexanoate

Purity: 99% CAS No.: 6243-10-3 Molecular Formula: C12H22O2 Formula Weight: 198.31 MDL No.: MFCD00046356

Formel: C₁₂H₂₂O₂
Molecular Weight: 198,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00046356
CAS-Nummer: 6243-10-3
EINECS: 228-354-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR