Nε-Boc-L-lysin ≥97%
Lieferant: Thermo Scientific
Synonyme:
Nε-Boc-L-lysine, H-Lys(Boc)-OH
B21738.03
B21738.06
B21738.03EA
79.9
EUR
B21738.03
B21738.06
Nε-Boc-L-lysin ≥97%
Nε-Boc-L-lysin
Purity: 97%
CAS No.: 2418-95-3
Molecular Formula: C11H22N2O4
Formula Weight: 246.31
Melting Point: ca 250° dec.
Beilstein Reference: 4252546
MDL No.: MFCD00037221
Formel:
C₁₁H₂₂N₂O₄ Molecular Weight: 246,31 g/mol Schmelzpunkt: ∼ 250 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037221 CAS-Nummer: 2418-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...