2-(4-Methyl-1,3-thiazol-5-yl)ethyl-acetat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Acetic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Acetoxyethyl)-4-methylthiazole, 4-Methyl-5-thiazolylethyl acetate

B21867.06 B21867.14 B21867.22
B21867.06EA 27.6 EUR
B21867.06 B21867.14 B21867.22
2-(4-Methyl-1,3-thiazol-5-yl)ethyl-acetat ≥98%
2-(4-Methyl-5-thiazolyl)ethyl acetate

Purity: 98+% CAS No.: 656-53-1 Molecular Formula: C8H11NO2S Formula Weight: 185.24 Flash Point: 110°(230°F) Refractive Index: 1.5100 Boiling Point: 116-118°/6mm MDL No.: MFCD00005338

Formel: C₈H₁₁NO₂S
Molecular Weight: 185,24 g/mol
Siedepunkt: 116…118 °C (6 mmHg)
Dichte: 1,147
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005338
CAS-Nummer: 656-53-1
EINECS: 211-515-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR