3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Phenazopyridine hydrochloride, 3-Phenylazo-2,6-diaminopyridine hydrochloride

B21886.06 B21886.14
B21886.06EA 53.8 EUR
B21886.06 B21886.14
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥98%
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride

Purity: 98+% CAS No.: 136-40-3 Molecular Formula: C11H11N5.HCl Formula Weight: 249.70 Melting Point: ca 240° dec. MDL No.: MFCD00035347

Formel: C₁₁H₁₂ClN₅
Molecular Weight: 249,70 g/mol
Schmelzpunkt: ∼ 240 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00035347
CAS-Nummer: 136-40-3
EINECS: 205-243-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR