N-Boc-D-glutamic acid 1-benzyl ester 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-Benzyl N-Boc-D-glutamate, Boc-D-Glu-OBzl, N-Boc-D-glutamic Acid 1-Benzyl Ester, N-(tert-Butoxycarbonyl)-D-glutamic Acid 1-Benzyl Ester
B21944.03
B21944.06
B21944.03EA
33
EUR
B21944.03
B21944.06
N-Boc-D-glutamic acid 1-benzyl ester 99%
N-Boc-D-glutamic acid 1-benzyl ester
Purity: 99%
CAS No.: 34404-30-3
Molecular Formula: C17H23NO6
Formula Weight: 337.37
MDL No.: MFCD00038266
Formel:
C₁₇H₂₃NO₆ Molecular Weight: 337.37 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038266 CAS-Nummer: 34404-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...