D(+)-Melezitose Hydrat ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol xhydrate

B22209.09 B22209.18
B22209.09EA 49.9 EUR
B22209.09 B22209.18
D(+)-Melezitose Hydrat ≥99%
D-(+)-Melezitose hydrate

Purity: 99% CAS No.: 207511-10-2 Molecular Formula: C18H32O16.xH2O Formula Weight: 504.46 (anhy) Melting Point: ca 160° dec. Beilstein Reference: 99539 Optical Rotation: +88°(c=4 in water) MDL No.: MFCD00149448

Formel: C₁₈H₃₄O₁₇
Molecular Weight: 522.45 g/mol
Schmelzpunkt: ∼ 160 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149448
CAS-Nummer: 207511-10-2
EINECS: 209-894-9
Merck Index: 14,5819

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR