L-Aspartic acid monosodium salt monohydrate 99%
Lieferant: Thermo Fisher Scientific
Synonyme: Sodium (S)-3-amino-3-carboxypropanoate hydrate, Sodium L-aspartate monohydrate, L-Aspartic acid sodium salt monohydrate
B22321.22 B22321.36
B22321.22EA 43.9 EUR
B22321.22 B22321.36
L-Aspartic acid monosodium salt monohydrate 99%
L-Aspartic acid monosodium salt monohydrate
Purity: 99% CAS No.: 323194-76-9 Molecular Formula: C4H6NNaO4.H2O Formula Weight: 173.11 (155.10anhy) MDL No.: MFCD00152960
Formel: C₄H₆NNaO₄·H₂O
Molecular Weight: 173.11 g/mol (155.10 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00152960
CAS-Nummer: 323194-76-9
EINECS: 223-264-0