Boc-Glu(OtBu)-OH 99%

Lieferant: Thermo Scientific

B22322.03 B22322.06
B22322.03EA 49.3 EUR
B22322.03 B22322.06
Boc-Glu(OtBu)-OH 99%
Boc-Glu(OtBu)-OH

Purity: 99% CAS No.: 13726-84-6 Molecular Formula: C14H25NO6 Formula Weight: 303.36 Melting Point: 110-112° MDL No.: MFCD00038257

Formel: C₁₄H₂₅NO₆
Molecular Weight: 303,355 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038257
CAS-Nummer: 13726-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR