Azo rubine
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Carmoisine B, acid Red 14, Brilliant Crimson Red, Sodium 4-hydroxy-3-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate
B22328.09
B22328.18
B22328.30
B22328.09EA
39.2
EUR
B22328.09
B22328.18
B22328.30
Azo rubine
Azo rubine
CAS No.: 3567-69-9
Molecular Formula: C20H12N2Na2O7S2
Formula Weight: 502.42
MDL No.: MFCD00003978
Formel:
C₂₀H₁₄N₂O₇S₂ Molecular Weight: 502.42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003978 CAS-Nummer: 3567-69-9 EINECS: 222-657-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...