O-Benzyl-L-serine 99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Ser(Bzl)-OH

B22393.06 B22393.14
B22393.06EA 104 EUR
B22393.06 B22393.14
O-Benzyl-L-serine 99%
O-Benzyl-L-serine

Purity: 99% CAS No.: 4726-96-9 Molecular Formula: C10H13NO3 Formula Weight: 195.22 Melting Point: 213° dec. Beilstein Reference: 2114846 Optical Rotation: +7° (c=1 in 1N HCl) MDL No.: MFCD00065937

Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,22 g/mol
Schmelzpunkt: 213 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00065937
CAS-Nummer: 4726-96-9
EINECS: 225-220-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR