N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98%
Lieferant: Thermo Scientific
Synonyme:
Boc-GABA-OH, N-Boc-γ-aminobutyric acid, 4-(Boc-amino)butyric Acid, 4-(tert-Butoxycarbonylamino)butyric Acid, N-Boc-4-aminobutyric Acid, N-BOC-gamma-Aminobutyric acid
B22407.03
B22407.06
B22407.03EA
34.7
EUR
B22407.03
B22407.06
N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98%
N-(tert-Butoxycarbonyl)-4-aminobutyric acid
Purity: 98+%
CAS No.: 57294-38-9
Molecular Formula: (CH3)3COCONH(CH2)3CO2H
Formula Weight: 203.24
Melting Point: 57-60°
MDL No.: MFCD00037313
Formel:
C₉H₁₇NO₄ Molecular Weight: 203,24 g/mol Schmelzpunkt: 57…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037313 CAS-Nummer: 57294-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...