2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 97%
Lieferant: Thermo Scientific
Synonyme: 2-Boc-thio-4,6-dimethylpyrimidine, O-tert-Butyl S-(4,6-dimethylpyrimidin-2-yl) carbonothioate
B22435.06 B22435.14
B22435.06EA 20.9 EUR
B22435.06 B22435.14
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 97%
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine
Purity: 97% CAS No.: 41840-28-2 Molecular Formula: C11H16N2O2S Formula Weight: 240.33 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 48-51° Beilstein Reference: 396984 MDL No.: MFCD00006080
Formel: C₁₁H₁₆N₂O₂S
Molecular Weight: 240,32 g/mol
Schmelzpunkt: 48…51 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00006080
CAS-Nummer: 41840-28-2
EINECS: 255-562-1