Sodium-1-naphthalenesulphonate, (max. 2% H₂O) 99% (Trockengewicht)

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Naphthalenesulphonic acid sodium salt, Naphthalene-1-sulphonic acid sodium salt

B22488.06 B22488.14 B22488.22
B22488.06EA 28.2 EUR
B22488.06 B22488.14 B22488.22
Sodium-1-naphthalenesulphonate, (max. 2% H₂O) 99% (Trockengewicht)
Sodium-1-naphthalenesulphonate

Purity: 99% CAS No.: 130-14-3 Molecular Formula: C10H7NaO3S Formula Weight: 230.22 Melting Point: 299-301° Beilstein Reference: 3639038 MDL No.: MFCD00064964

Formel: C₁₀H₈O₃S
Molecular Weight: 230,21 g/mol
Schmelzpunkt: 299…301 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064964
CAS-Nummer: 130-14-3
EINECS: 204-976-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR