5-Methoxy-2-benzimidazolinone 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
5-Methoxy-1H-benzo[d]imidazol-2(3H)-one, 2-Hydroxy-5-methoxybenzimidazole
B22580.03
B22580.06
B22580.03EA
39.1
EUR
B22580.03
B22580.06
5-Methoxy-2-benzimidazolinone 98%
5-Methoxy-2-benzimidazolinone
Purity: 98%
CAS No.: 2080-75-3
Molecular Formula: C8H8N2O2
Formula Weight: 164.17
Melting Point: ca 255° dec.
Beilstein Reference: 141043
MDL No.: MFCD00051946
Formel:
C₈H₈N₂O₂ Schmelzpunkt: ∼ 255 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051946 CAS-Nummer: 2080-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...