4-Cyano-4''-n-pentyl-p-terphenyl 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4''-Pentyl-[1,1':4',1''-terphenyl]-4-carbonitrile, 4-Cyano-4''-pentyl-p-terphenyl
B22607.02
B22607.88
B22607.02EA
65.6
EUR
B22607.02
B22607.88
4-Cyano-4''-n-pentyl-p-terphenyl 99%
4-Cyano-4''-n-pentyl-p-terphenyl
Purity: 99%
CAS No.: 54211-46-0
Molecular Formula: C24H23N
Formula Weight: 325.45
Melting Point: 128-130°
UN No.: UN3439
MDL No.: MFCD00799423
Formel:
C₂₄H₂₃N Molecular Weight: 325.45 g/mol Schmelzpunkt: 128…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00799423 CAS-Nummer: 54211-46-0 EINECS: 259-028-9 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...