N-Carbobenzoxy-L-serine benzyl ester 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Cbz-L-serine Benzyl Ester, Z-Ser-OBzl, Benzyl ((benzyloxy)carbonyl)-L-serinate
B22647.03
B22647.06
B22647.03EA
37.5
EUR
B22647.03
B22647.06
N-Carbobenzoxy-L-serine benzyl ester 99%
N-Carbobenzoxy-L-serine benzyl ester
Purity: 99%
CAS No.: 21209-51-8
Molecular Formula: C18H19NO5
Formula Weight: 329.32
Melting Point: 82-84°
MDL No.: MFCD00037824
Formel:
C₁₈H₁₉NO₅ Molecular Weight: 329.35 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037824 CAS-Nummer: 21209-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...