Phloroglucide 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: PHBF, 2,3',4,5',6-Pentahydroxybiphenyl

B22693.06 B22693.14
B22693.06EA 278 EUR
B22693.06 B22693.14
Phloroglucide 95%
Phloroglucide

Purity: 95% CAS No.: 491-45-2 Molecular Formula: C12H10O5 Formula Weight: 234.21 Melting Point: ca 296° dec. Beilstein Reference: 3324960 MDL No.: MFCD00044547

Formel: C₁₂H₁₀O₅
Schmelzpunkt: ∼ 296 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00044547
CAS-Nummer: 491-45-2

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Cream to pale pink or pale brown
Form powder
Assay (HPLC) >94.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR