N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-D-alanin ≥98%

Lieferant: Thermo Scientific

Synonyme: N-Boc-D-alanine, Boc-D-Ala-OH

B22706.06 B22706.14
B22706.06EA 39.9 EUR
B22706.06 B22706.14
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-D-alanin ≥98%
Boc-D-Alanine

Purity: 98+% CAS No.: 7764-95-6 Molecular Formula: C8H15NO4 Formula Weight: 189.21 Melting Point: 81-84° Beilstein Reference: 2048396 Optical Rotation: +26° (c=2 in acetic acid) MDL No.: MFCD00063123

Formel: C₈H₁₅NO₄
Molecular Weight: 189,2 g/mol
Schmelzpunkt: 81…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063123
CAS-Nummer: 7764-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR