5-Chlor-2-thiophencarbaldehyd 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chloro-5-thiophenecarboxaldehyde, 2-Chloro-5-formylthiophene

B22833.03 B22833.06 B22833.14
B22833.03EA 39 EUR
B22833.03 B22833.06 B22833.14
5-Chlor-2-thiophencarbaldehyd 97%
5-Chlor-2-thiophencarbaldehyd

Purity: 97% CAS No.: 7283-96-7 Molecular Formula: C5H3ClOS Formula Weight: 46.60 Storage Temperature: 2°C to 8°C Flash Point: 97°(208°F) Refractive Index: 1.6040 Boiling Point: 88-90°/9mm MDL No.: MFCD00047090

Formel: C₅H₃ClOS
Molecular Weight: 146,59 g/mol
Siedepunkt: 88…90 °C (9 mmHg)
Dichte: 1,376
Flammpunkt: 97 °C (208 °F)
MDL: MFCD00047090
CAS-Nummer: 7283-96-7
EINECS: 230-708-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR