N-[Tris(hydroxymethyl)methyl]acrylamide 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(1,3-Dihydroxy-2-(hydroxymethyl)propan-2-yl)acrylamide

B22854.06 B22854.14 B22854.22
B22854.06EA 62.4 EUR
B22854.06 B22854.14 B22854.22
N-[Tris(hydroxymethyl)methyl]acrylamide 99%
N-[Tris(hydroxymethyl)methyl]acrylamide

Purity: 99% CAS No.: 13880-05-2 Molecular Formula: C7H13NO4 Formula Weight: 175.18 Storage Temperature: 2°C to 8°C Melting Point: 134-138° MDL No.: MFCD00081045

Formel: C₇H₁₃NO₄
Molecular Weight: 175,18 g/mol
Schmelzpunkt: 134…138 °C
MDL: MFCD00081045
CAS-Nummer: 13880-05-2
EINECS: 237-645-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR