N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine 98%

Lieferant: Thermo Scientific

Synonyme: N-Boc-O-benzyl-L-tyrosine, BOC-O-Benzyl-L-tyrosine, Boc-Tyr(Bzl)-OH

B23455.03 B23455.06 B23455.14
B23455.03EA 22.6 EUR
B23455.03 B23455.06 B23455.14
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine 98%
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine

Purity: 98% CAS No.: 2130-96-3 Molecular Formula: C21H25NO5 Formula Weight: 371.43 Storage Temperature: 2°C to 8°C Melting Point: 108-110° MDL No.: MFCD00065597

Formel: C₂₁H₂₅NO₅
Molecular Weight: 371,43 g/mol
Schmelzpunkt: 108…110 °C
MDL: MFCD00065597
CAS-Nummer: 2130-96-3
EINECS: 218-349-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR