ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole] 98%
Lieferant: Thermo Scientific
Synonyme:
ABD-F, 4-Fluoro-7-sulfobenzofurazan, ammonium salt, 7-Fluoro-2,1,3-Benzoxadiazole-4-sulphonamide, 4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole, 4-(Aminosulfonyl)-7-fluorobenzofurazane, 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonamide, 7-Fluorobenzofurazan-4-sulfonamide
B23586.MB
B23586.MC
B23586.MBEA
391
EUR
B23586.MB
B23586.MC
ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole] 98%
ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole]
Purity: 98%
CAS No.: 91366-65-3
Molecular Formula: C6H4FN3O3S
Formula Weight: 217.17
Melting Point: 143-147°
MDL No.: MFCD00036687
Formel:
C₆H₄FN₃O₃S Molecular Weight: 217,17 g/mol Schmelzpunkt: 143…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036687 CAS-Nummer: 91366-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...