Zinc propionate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23636.0B
B23636.18
B23636.30
B23636.0BEA
403
EUR
B23636.0B
B23636.18
B23636.30
Zinc propionate 97%
Zinc propionate
Purity: 97%
CAS No.: 557-28-8
Molecular Formula: (CH3CH2CO2)2Zn
Formula Weight: 211.53
MDL No.: MFCD00045937
Formel:
(CH₃CH₂CO₂)₂Zn Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045937 CAS-Nummer: 557-28-8 EINECS: 209-167-6 Merck Index: 14,10153 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...