N-Acetyl-D-phenylalanin 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ac-Phe-OH
B23812.03
B23812.06
B23812.14
B23812.03EA
19.7
EUR
B23812.03
B23812.06
B23812.14
N-Acetyl-D-phenylalanin 99%
N-Acetyl-L-phenylalanine
Purity: 99%
CAS No.: 2018-61-3
Molecular Formula: C11H13NO3
Formula Weight: 207.23
Melting Point: 166-170°
Optical Rotation: +40° (c=1 in methanol)
MDL No.: MFCD00063158
Formel:
C₁₁H₁₃NO₃ Molecular Weight: 207.2 g/mol Schmelzpunkt: 166…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063158 CAS-Nummer: 2018-61-3 EINECS: 217-959-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...