Iron(II) fumarate 94%

Lieferant: Thermo Scientific

Synonyme: Ferrous fumarate

B23956.0B B23956.30
B23956.0BEA 112 EUR
B23956.0B B23956.30
Iron(II) fumarate 94%
Iron(II) fumarate

Purity: 94% CAS No.: 141-01-5 Molecular Formula: C4H2FeO4 Formula Weight: 169.90 Melting Point: 300° MDL No.: MFCD00058315

Formel: C₄H₂FeO₄
Molecular Weight: 169,90 g/mol
Schmelzpunkt: >300 °C
Dichte: 2,44
Lagertemperatur: Raumtemperatur
MDL: MFCD00058315
CAS-Nummer: 141-01-5
EINECS: 205-447-7
Merck Index: 14,4046

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR