5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
Lieferant: Thermo Scientific
Synonyme:
5(4)-Amino-4(5)-imidazolecarboxamide Hydrochloride, AICA, 5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 4-Amino-5-imidazolecarboxamide hydrochloride, 5-Amino-4-imidazolecarboxamide hydrochloride
B24012.06
B24012.14
B24012.06EA
70.8
EUR
B24012.06
B24012.14
5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride
Purity: 98%
CAS No.: 72-40-2
Molecular Formula: C4H6N4O.HCl
Formula Weight: 162.58
Odour: Odourless
Melting Point: ca 270° dec.
Beilstein Reference: 3701645
MDL No.: MFCD00012704
Formel:
C₄H₇ClN₄O Molecular Weight: 162,58 g/mol Schmelzpunkt: ∼ 270 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012704 CAS-Nummer: 72-40-2 EINECS: 200-778-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...