Ethyl-3-(dimethylamino)acrylat 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl 3-(dimethylamino)acrylate, 3-(Dimethylamino)acrylic Acid Ethyl Ester, Ethyl 3-(N,N-dimethylamino)acrylate
B24291.18
B24291.09
B24291.30
B24291.18EA
128
EUR
B24291.18
B24291.09
B24291.30
Ethyl-3-(dimethylamino)acrylat 99%
Ethyl-3-(dimethylamino)acrylate
Purity: 99%
CAS No.: 924-99-2
Molecular Formula: C7H13NO2
Formula Weight: 143.19
Refractive Index: 1.5120
Melting Point: 17-18°
Boiling Point: 118-121°/10mm
MDL No.: MFCD00144269
Formel:
C₇H₁₃NO₂ Molecular Weight: 143,19 g/mol Siedepunkt: 118…121 °C (10 mmHg) Schmelzpunkt: 17…18 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144269 CAS-Nummer: 924-99-2 EINECS: 402-650-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...