gamma-Phenyl-gamma-Butyrolacton 97%
Lieferant: Thermo Scientific
Synonyme:
5-Phenyltetrahydrofuran-2-one
B24364.06
B24364.14
B24364.06EA
35.5
EUR
B24364.06
B24364.14
gamma-Phenyl-gamma-Butyrolacton 97%
gamma-Phenyl-gamma-butyrolactone
Purity: 97%
CAS No.: 1008-76-0
Molecular Formula: C10H10O2
Formula Weight: 162.19
Flash Point:
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol Siedepunkt: 305 °C Schmelzpunkt: 35…38 °C Dichte: 1,155 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005398 CAS-Nummer: 1008-76-0 EINECS: 213-761-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...