D-Asparagine monohydrate 99%
Lieferant: Thermo Scientific
Synonyme:
(R)-2-Aminosuccinamic acid hydrate, D-2-Aminosuccinamic acid, H-D-Asn-OH·H₂O, (R)-2,4-Diamino-4-oxobutanoic acid hydrate
B24556.14
B24556.22
B24556.14EA
38
EUR
B24556.14
B24556.22
D-Asparagine monohydrate 99%
D-Asparagine monohydrate
Purity: 99%
CAS No.: 5794-24-1
Molecular Formula: C4H8N2O3.H2O
Formula Weight: 150.14 (132.12anhy)
Melting Point: 274-276° dec.
MDL No.: MFCD00149558
Formel:
C₄H₁₀N₂O₄ Molecular Weight: 150,14 g/mol (132,12 anhy) Schmelzpunkt: 274…276 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149558 CAS-Nummer: 5794-24-1 Merck Index: 14,837 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...