Benzyl-3-hydroxyphenylacetate 98%

Lieferant: Thermo Scientific

Synonyme: Benzyl 2-(3-hydroxyphenyl)acetate

B24679.06 B24679.14
B24679.06EA 73.7 EUR
B24679.06 B24679.14
Benzyl-3-hydroxyphenylacetate 98%
Benzyl-3-hydroxyphenylacetate

Purity: 98% CAS No.: 295320-25-1 Molecular Formula: C15H14O3 Formula Weight: 242.27 Melting Point: 79-81° MDL No.: MFCD04039768

Formel: C₁₅H₁₄O₃
Molecular Weight: 242,27 g/mol
Schmelzpunkt: 79…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039768
CAS-Nummer: 295320-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR