Methyl malonamate 98%

Lieferant: Thermo Scientific

Synonyme: Methyl 3-amino-3-oxopropanoate

B25326.14 B25326.22
B25326.14EA 130 EUR
B25326.14 B25326.22
Methyl malonamate 98%
Methyl malonamate

Purity: 98% CAS No.: 51513-29-2 Molecular Formula: C4H7NO3 Formula Weight: 117.11 Melting Point: 38-40° MDL No.: MFCD00674533

Formel: C₄H₇NO₃
Molecular Weight: 117,11 g/mol
Schmelzpunkt: 38…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00674533
CAS-Nummer: 51513-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR