Ethyl-1H-pyrazol-4-carboxylat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
ethyl 1H-pyrazole-4-carboxylate, Ethyl 4-pyrazolecarboxylate
B25351.06
B25351.14
B25351.06EA
138
EUR
B25351.06
B25351.14
Ethyl-1H-pyrazol-4-carboxylat 98%
Ethyl-4-pyrazolecarboxylate
Purity: 98%
CAS No.: 37622-90-5
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Melting Point: 75-80°
Boiling Point: 138-140°/3mm
MDL No.: MFCD00010844
Formel:
C₆H₈N₂O₂ Molecular Weight: 140,14 g/mol Siedepunkt: 138…140 °C (3 mmHg) Schmelzpunkt: 75…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010844 CAS-Nummer: 37622-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...