2-Bromo-4-nitrophenetole 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Bromo-4-ethoxynitrobenzene

B25433.22 B25433.06
B25433.22EA 0 EUR
B25433.22 B25433.06
2-Bromo-4-nitrophenetole 99%
2-Bromo-4-nitrophenetole

Purity: 99% CAS No.: 58244-42-1 Molecular Formula: C8H8BrNO3 Formula Weight: 246.06 Melting Point: 94-98° MDL No.: MFCD12547883

Formel: C₈H₈BrNO₃
Molecular Weight: 246,06 g/mol
Schmelzpunkt: 94…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12547883
CAS-Nummer: 58244-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR