Malonamamidine hydrochloride ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Malonamideamidine hydrochloride

B25569.14 B25569.22
B25569.14EA 75.4 EUR
B25569.14 B25569.22
Malonamamidine hydrochloride ≥98%
Malonamamidine hydrochloride

Purity: 98+% CAS No.: 34570-17-7 Molecular Formula: C3H7N3O.HCl Formula Weight: 137.57 Melting Point: 170-174° MDL No.: MFCD00013007

Formel: C₃H₇N₃O·HCl
Molecular Weight: 137,57 g/mol
Schmelzpunkt: 170…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013007
CAS-Nummer: 34570-17-7
EINECS: 252-095-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR