7-Bromo-1,2,3,4-tetrahydroisoquinoline 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B25712.03
B25712.06
B25712.03EA
71.3
EUR
B25712.03
B25712.06
7-Bromo-1,2,3,4-tetrahydroisoquinoline 97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline
Purity: 97%
CAS No.: 17680-55-6
Molecular Formula: C9H10BrN
Formula Weight: 212.09
Melting Point: 32-35°
MDL No.: MFCD06739047
Formel:
C₉H₁₀BrN Molecular Weight: 212.09 g/mol Schmelzpunkt: 32…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739047 CAS-Nummer: 17680-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...