DL-Cysteine hydrochloride ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
H-DL-Cys-OH·HCl, 2-Amino-3-mercaptopropanoic acid hydrochloride
H25779.22
H25779.06
H25779.22EA
798
EUR
H25779.22
H25779.06
DL-Cysteine hydrochloride ≥98%
DL-Cysteine hydrochloride
Purity: 98+%
CAS No.: 10318-18-0
Molecular Formula: C3H7NO2S.HCl
Formula Weight: 157.62
Storage Temperature: 2°C to 8°C
Melting Point: 140-142°
Beilstein Reference: 4150473
MDL No.: MFCD00064552
Formel:
C₃H₇NO₂S·HCl Molecular Weight: 157.62 g/mol Schmelzpunkt: 140…142 °C |
MDL:
MFCD00064552 CAS-Nummer: 10318-18-0 EINECS: 233-698-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...