(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
Lieferant: Thermo Scientific
Achtung
Synonyme: (S)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-BINAM, (S)-(-)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-1,1'-Bi(2-naphthylamine), (S)-(-)-2,2'-Diamino-1,1'-binaphthyl
H25784.MD H25784.03
H25784.MDEA 48.2 EUR
H25784.MD H25784.03
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene
Purity: 97% CAS No.: 18531-95-8 Molecular Formula: C20H16N2 Formula Weight: 284.36 Melting Point: 236-242° Beilstein Reference: 2419071 Optical Rotation: -150° (c=1 in pyridine) MDL No.: MFCD00145204
Formel: C₂₀H₁₆N₂
Molecular Weight: 284,362 g/mol
Schmelzpunkt: 236…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00145204
CAS-Nummer: 18531-95-8