(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 97%
Lieferant: Thermo Scientific
Achtung
Synonyme: (R)-(+)-2,2'-Diamino-1,1'-binaphthyl, (R)-(+)-1,1'-Binaphthyl-2,2'-diamine, (R)-(+)-BINAM, (R)-[1,1'-Binaphthalene]-2,2'-diamine
H25785.03 H25785.MD
H25785.03EA 152 EUR
H25785.03 H25785.MD
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 97%
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene
Purity: 97% CAS No.: 18741-85-0 Molecular Formula: C20H16N2 Formula Weight: 284.36 Melting Point: 242-243° Beilstein Reference: 5279621 Optical Rotation: +150° (c=1 in pyridine) MDL No.: MFCD00145204
Formel: C₂₀H₁₆N₂
Molecular Weight: 284,362 g/mol
Schmelzpunkt: 242…243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00145204
CAS-Nummer: 18741-85-0