4-n-Propylpiperidine 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Propylpiperidine
H25848.MD
H25848.03
H25848.MDEA
57.7
EUR
H25848.MD
H25848.03
4-n-Propylpiperidine 95%
4-n-Propylpiperidine
Purity: 95%
CAS No.: 22398-09-0
Molecular Formula: C8H17N
Formula Weight: 127.23
Flash Point: 48°(118°F)
Refractive Index: 1.4530
Odour: Amine-like
Boiling Point: 178-180°
UN No.: UN2733
MDL No.: MFCD00023151
Formel:
C₈H₁₇N Molecular Weight: 127,23 g/mol Siedepunkt: 178…180 °C Dichte: 0,864 Flammpunkt: 48 °C (118 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023151 CAS-Nummer: 22398-09-0 EINECS: 244-956-9 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...