(S)-2-(Trifluoromethyl)pyrrolidine 95%

Lieferant: Thermo Fisher Scientific
Gefahr

H25854.MD H25854.03
H25854.MDEA 37.3 EUR
H25854.MD H25854.03
(S)-2-(Trifluoromethyl)pyrrolidine 95%
(S)-2-(Trifluoromethyl)pyrrolidine

Purity: 95% CAS No.: 119580-41-5 Molecular Formula: C5H8F3N Formula Weight: 139.12 Flash Point: 26°(79°F) Melting Point: 27-31° Boiling Point: 110-112° Optical Rotation: +10° (c=1 in methanol) UN No.: UN2925 MDL No.: MFCD03093766

Formel: C₅H₈F₃N
Molecular Weight: 139.12 g/mol
Siedepunkt: 110…112 °C
Schmelzpunkt: 27…31 °C
Flammpunkt: 26 °C (79 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03093766
CAS-Nummer: 119580-41-5
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR