3-Amino-4-phenyl-1H-pyrazole 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Phenyl-1H-pyrazol-3-amine
H25895.06
H25895.03
H25895.06EA
143
EUR
H25895.06
H25895.03
3-Amino-4-phenyl-1H-pyrazole 95%
3-Amino-4-phenyl-1H-pyrazole
Purity: 95%
CAS No.: 5591-70-8
Molecular Formula: C9H9N3
Formula Weight: 159.19
Melting Point: 170-173°
MDL No.: MFCD01693715
Formel:
C₉H₉N₃ Molecular Weight: 159.19 g/mol Schmelzpunkt: 170…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01693715 CAS-Nummer: 5591-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...