D-Phenylalanin-methylester Hydrochlorid 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl
H25920.14
H25920.06
H25920.14EA
150
EUR
H25920.14
H25920.06
D-Phenylalanin-methylester Hydrochlorid 99%
D-Phenylalanine methyl ester hydrochloride
Purity: 99%
CAS No.: 13033-84-6
Molecular Formula: C10H13NO2.HCl
Formula Weight: 215.68
Melting Point: 158-162°
MDL No.: MFCD00066112
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 215,68 g/mol Schmelzpunkt: 158…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066112 CAS-Nummer: 13033-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...