5-Formyl-4-methylthiophene-2-boronic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (5-Formyl-4-methylthiophen-2-yl)boronic acid

H25947.06 H25947.03
H25947.06EA 224 EUR
H25947.06 H25947.03
5-Formyl-4-methylthiophene-2-boronic acid 97%
5-Formyl-4-methylthiophene-2-boronic acid

Purity: 97% CAS No.: 352530-25-7 Molecular Formula: C6H7BO3S Formula Weight: 169.95 Melting Point: 118-120° MDL No.: MFCD03093890

Formel: C₆H₇BO₃S
Molecular Weight: 169,95 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093890
CAS-Nummer: 352530-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR